Would you like to log in again?
Slit Seal, BioChromato
Supplier: BioChromato
R80.120.10 R80.120.03 R80.120.00
102962-944BX 4286.15 USD InStock
102962-944 102962-940 102962-942
Slit Seal, BioChromato
Seals Microplate Sealing Films and Foils
The Slit Seal is a resistant, instant self-closing 96-well plate seal
  • Self-closes instantly
  • No cross-contamination between wells
  • Allows next day analysis
  • No adhesive on well spots
  • Pipette tips insert easily with less friction
The adhesive-free, pre-cut slits are made of silicone and PET, and allow easy insertion and withdrawal of pipette tips and sampling needles without dragging. The Slit Seal’s main purpose is to prevent solvent evaporation and cross-contamination.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty