Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Suppliers found in search results
    Sort by:

  • Search Within Results

You Searched for: "led-transilluminators"

https://us.vwr.com/stibo/smallweb/std.lang.all/94/59/18989459.jpg

LED transilluminators provide offer superior excitation with bright LED lights that are safer than UV and can be combined with a number of different filters to protect the eyes during gel cutting. These transilluminators do not cause burn to the skin and eyes, and intensity is adjustable over a range of brilliance. Transilluminators provide excellent uniformity across the full filter area. Higher sensitivity leads to compatibility with most stains.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
Sort by:
 
 
 
 

Catalog Number: (10001-870)
Supplier:  G-Biosciences
Description:   - Compact (30 mm in height) and lightweight, it is convenient to move around
- User friendly, it is easy to view and cut gel - No need to wear am...
Supplier:  G-Biosciences
Description:   The Dual wavelength LED Transilluminator has high uniformity across the whole viewing area.
Supplier:  G-Biosciences
Description:   Safer to replace UV, this LED transilluminator provides strong R/G/B LEDs for excitation. Triple wavelengths are embedded in the same transilluminator...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 3  of 3