Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-E2F3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AY4
10803-026EA 423.98 USD InStock
10803-026
Anti-E2F3 Rabbit Polyclonal Antibody
Antibodies
The E2F transcription factor 3 (E2F3) is a member of a small family of transcription factors that function through binding of DP interaction partner proteins. E2F3 recognizes a specific sequence motif in DNA and interacts directly with the retinoblastoma protein (pRB) to regulate the expression of genes involved in the cell cycle (1). Like the related E2F1 and E2F2, E2F3 is essential for cellular proliferation and progression through the cell cycle (2). Altered copy number and activity of this gene have been observed in a number of human cancers (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 μg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: E2F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty