Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DPF3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AX1
10802-542EA 423.98 USD InStock
10802-542
Anti-DPF3 Rabbit Polyclonal Antibody
Antibodies
The DPF3 protein, also known as Cerd4, is a member of the d4 gene family of transcription modulators that also includes DPF1/Neud4 and DPF2/Requiem (1). DPF3 has been shown to be a epigenetic key factor for heart and muscle development and can bind to methylated and acetylated lysine residues of histone 3 and 4, suggesting that DPF3 may play a role in recruiting chromatin remodeling complexes to acetylated histones (2). Two isoforms of DPF3, DPF3a and DPF3b, are required as transcriptional co-activators in SWI/SNF complex-dependent activation of the NF-kappaB RelA/p50 heterodimer (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 μg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DPF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty