Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DPAGT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AX0
10802-540EA 423.98 USD InStock
10802-540
Anti-DPAGT1 Rabbit Polyclonal Antibody
Antibodies
The UDP-N-acetylglucosamine-dolichyl-phosphate N-acetyl-glucosaminephosphotransferase (DPAGT1) is an enzyme that catalyzes the first step in the dolichol-linked oligosaccharide pathway for glycoprotein biosynthesis. Mutations in this integral endoplasmic reticulum (ER) membrane protein enzyme belongs to the glycosyltransferase family 4 results in the congenital disorder of glycosylation type Ij with symptoms such as severe hypotonia, medically intractable seizures, mental retardation, microcephaly, and exotropia. Recent experiments have shown that DPAGT1 is a target of the Wnt/beta-catenin signaling pathway, with Wnt3a inducing higher DPAGT1 mRNA expression.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 2.5 μg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DPAGT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty