Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DOCK8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AW8
10801-170EA 423.98 USD InStock
10801-170
Anti-DOCK8 Rabbit Polyclonal Antibody
Antibodies
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors (1). DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse (reviewed in 2). Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DOCK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty