Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DISP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AW1
10801-144EA 423.98 USD InStock
10801-144
Anti-DISP2 Rabbit Polyclonal Antibody
Antibodies
Disp2 is the second of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored Hedgehog from producing cells. Hedgehog is a major player in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis and the Disp proteins have been implicated in these pathways. Recently, it has been shown that Disp2 is translationally regulated by the microRNA miR-214 in zebrafish. Expression of this miRNA decreased Disp2 promoter activity in vitro and its overexpression in zebrafish resulted in a phenotype identical to that observed by Disp2 mutants. At least two isoforms of Disp2 are known to exist. This Disp2 antibody is predicted to not cross-react with Disp1 or Disp3.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DISP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty