Anti-DEPTOR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AV0
10802-438EA 518.29 USD
10802-438
Anti-DEPTOR Rabbit Polyclonal Antibody
Antibodies
DEPTOR is an mTOR (mammalian target of rapamycin)-interacting protein whose expression is negatively regualted by mTOR complex 1 (mTORC1) and mTORC2 (1). Loss of DEPTOR activates RP6SK1, Akt1, and SGK1, promotes cell growth and survival, and activates mTORC1 and mTORC2 kinase activities. DEPTOR expression is low in most cancers, but is often elevated in multiple myelomas (1). In respons to stimulation by mitogens, DEPTOR is rapidly phosphorylated leading to its ubiquitination and degradation, allowing mTOR to generate an auto-amplification loop to promotes its own full activation (2).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DEPTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR