Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DDC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AU2
10801-114EA 423.98 USD InStock
10801-114
Anti-DDC Rabbit Polyclonal Antibody
Antibodies
DOPA decarboxylase (DDC) belongs to the group II decarboxylase family of proteins (1). It is an important protein in the catecholamine biosynthesis pathway. DDC catalyzes the second reaction in the biosynthesis of catecholamines, trace amines and serotonin (1,2). It can form a homodimer and is expressed in the central nervous system (2). DDC can be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease, neuroendocrine tumors and drug abuse (3,4). Defects in DDC gene may cause the autosomal recessive disorder AADC deficiency (4).

Recommended Dilutions: ELISA: User Optimized; Immunohistochemsitry: User Optimized; Immunofluorescence Microscopy: User Optimized; Western Blot: User Optimized; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty