Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DDA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AU1
10801-112EA 423.98 USD InStock
10801-112
Anti-DDA1 Rabbit Polyclonal Antibody
Antibodies
DDA1 (DET1 and DDB1 associated 1), along with DET1 and DDB1 and a member of the UBE2E group of canonical ubiquitin-conjugating enzymes, comprise DDD-E2 complexes, which interact with multiple ubiquitin E3 ligases. One of these E3 ligases is Cul4-containing E3 ligase complex CRL4. Cells depleted of DDA1 spontaneously accumulate double-stranded DNA breaks in a similar fashion as Cul4A-, Cul4B-, or WDR23-depleted cells, suggesting that DDA1 interacts with the CRL4 complex and may be involved in the ubiquitination and subsequent proteasomal degradation of target proteins.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 μg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DDA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty