Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DCANP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AT3
10800-988EA 423.98 USD InStock
10800-988
Anti-DCANP1 Rabbit Polyclonal Antibody
Antibodies
The DCNP1 (dendritic cell nuclear protein 1) gene is specifically expressed in dendritic cells, potent antigen-presenting cells involved in activating naive T cells to initiate antigen-specific immune response (1). It is localized mainly in the perinucleus. One of the alleles (A/T) of this gene causes premature translation termination at aa 117 and has been associated with an increased prevalence of major depression in humans (2,3). DCNP1 may play a role in the pathogenesis of depressive disorders by enhancing corticotropin-releasing hormone expression in the hypothalamic paraventricular nucleus (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DCANP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty