Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DAP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AS4
10801-088EA 423.98 USD InStock
10801-088
Anti-DAP Rabbit Polyclonal Antibody
Antibodies
The Death-associated protein (DAP) is a basic proline-rich 15kDa protein that as a positive mediator of programmed cell death that is induced by interferon-gamma (1). DAP is also a direct substrate of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Under rich nutrient conditions, mTOR phosphorylates DAP at Ser3 and Ser51; under starvation conditions, these residues are dephosphorylated and DAP is converted into an active suppressor of autophagy (2).

  • Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: User Optimized; Immunofluorescence Microscopy: User Optimized; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty