Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCNO Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AR9
10801-050EA 423.98 USD InStock
10801-050
Anti-CCNO Rabbit Polyclonal Antibody
Antibodies
Cyclin O, also known as CCNO, has recently been identified as a Cdk1- and Cdk2-activating cyclin specific to apoptosis in lymphoid cells. Cyclin O binds to and activates Cdk2 in response to instrinsic apoptotic stimuli such as glucocorticoids or DNA-damaging agents. Supression of Cyclin O expression by shRNA leads to the inhibition of glucocorticoid and DNA-damage-induced apoptosis due to a failure of apical caspase activation while leaving the CD95 death receptor-mediated apoptosis intact. Note: this gene, which had a previous symbol of UNG2, was erroneously identified as a uracil DNA glycosylase. A later publication identified this gene's product as a cyclin protein family member.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CCNO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty