Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CXCR3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AR4
10800-968EA 423.98 USD InStock
10800-968
Anti-CXCR3 Rabbit Polyclonal Antibody
Antibodies
The CXCR3 protein is a G protein-coupled receptor with selectivity for three chemokines, termed CXCL9/Mig (monokine induced by interferon-gamma), CXCL10/IP10 (interferon-gamma-inducible 10 kDa protein) and CXCL11/I-TAC (interferon-inducible T cell alpha-chemoattractant) (reviewed in 1). Binding of chemokines to this protein induces cellular responses that are involved in leukocyte traffic, most notably integrin activation, cytoskeletal changes and chemotactic migration (2). Similar to other chemokine receptors such as CXCR4, CXCR3 has been implicated in several malignancies as a biomarker of tumor behavior as well as a potential therapeutic target (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 μg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CXCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty