Anti-CX3CL1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AR2
10802-428EA 518.29 USD
10802-428
Anti-CX3CL1 Rabbit Polyclonal Antibody
Antibodies
Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-a in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.

  • Recommended Dilutions: ELISA: 1:5000-1:20,000; Immunoprecipitation: User Optimized; Western Blot: 0.5-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CX3CL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR