Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTHRC1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AP3
10800-934EA 423.98 USD InStock
10800-934
Anti-CTHRC1 Rabbit Polyclonal Antibody
Antibodies
The Collagen triple helix repeat-containing protein (CTHRC1) is a glycosylated, secreted protein that is transiently expressed in the arterial wall in response to injury where it is thought to contribute to vascular remodeling by inhibiting collagen expression and deposition and promoting cell migration (1). CTHRC1 is highly expressed multiple human cancers, including pancreatic cancer where it plays a significant role in the progression and metastasis of the disease by regulating migration and adhesion of tumors cells (2). CTHRC1 overexpression can also be used as an independent prognostic marker in gastric cancer (3). Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CTHRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty