Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CORO7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AM6
10800-886EA 423.98 USD InStock
10800-886
Anti-CORO7 Rabbit Polyclonal Antibody
Antibodies
Coronins, such as Coronin 7, constitute an evolutionarily conserved family of WD-repeat actin-binding proteins whose function is thought to be regulating the cytoskeleton. Unlike other cytoplasmically localized Coronins, Coronin 7 localizes in part to the Golgi complex and is thought to play a role in Golgi complex morphology and function. Other experiments indicate that Coronin 7 interacts with the anti-proliferative protein TOB1 and enhances its degradation though the SCF ubiquitin ligase (E3) complex.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 2.5 µg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CORO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty