Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNRIP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AM3
10802-416EA 423.98 USD InStock
10802-416
Anti-CNRIP1 Rabbit Polyclonal Antibody
Antibodies
The CNRIP1 (cannabinoid receptor interacting protein 1) protein is a G-protein coupled receptor which interacts with the C-terminal tail of cannabinoid receptor 1 (CB1) and is thought to play a role in synaptic plasticity, analgesia, appetite, and neuroprotection. One isoform of CNRIP1, CNRIP1a, modulates the constitutive CB1 receptor activity in the central nervous system (CNS), while the role of the shorter isoform CNRIP1b is yet unknown. CNRIP1 has been suggested as a potential target for CNS drug discovery.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: User Optimized; Immunofluorescence Microscopy: User Optimized; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CNRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty