Anti-CNOT4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AM2
10802-414EA 518.29 USD
10802-414
Anti-CNOT4 Rabbit Polyclonal Antibody
Antibodies
CNOT4 is a component of the CCR4-NOT transcription complex, a complex that is implicated in the repression of RNA polymerase II transcription. In the CCR4-NOT complex, CNOT4 acts as an E3 ubiquitin-protein ligase and interacts with a subset of E2 ubiquitin-conjugating enzymes through a unique C4C4 RING domain. This E3 ligase activity was shown to be dependent on the selective and specific interaction with the ubiquitin conjugating enzyme UbcH5B. In yeast, mutations in CNOT4 that prevented its interaction with the UbcH5B homolog UBC4 caused increased sensitivity to hydroxyurea, heat shock, and hygromycin B, suggesting that CNOT4 and UbcH5B are involved in stress response in vivo. Multiple isoforms of CNOT4 are known to exist.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CNOT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR