Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CIDEA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AK3
10800-820EA 423.98 USD InStock
10800-820
Anti-CIDEA Rabbit Polyclonal Antibody
Antibodies
Apoptosis is related to many diseases and induced by a family of cell death receptors. Cell death signals are transduced by DD-, DED-, or CARD-containing molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase DFF40/CAD, which is chaperoned and inhibited by DFF45/ICAD. DFF45 related proteins CIDE-A and CIDE-B (for cell death-inducing DFF-like effector A and B) were recently identified. CIDE contains a new type of domain termed CIDE-N, which has high homology with the regulatory domains of DFF45/ICAD and DFF40/CAD. Expression of CIDE-A induces DNA fragmentation and activates apoptosis, which is inhibited by DFF45. CIDE-A is expressed in many tissues.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CIDEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty