Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDKN1C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AJ7
10800-746EA 423.98 USD InStock
10800-746
Anti-CDKN1C Rabbit Polyclonal Antibody
Antibodies
The cyclin-dependent kinase inhibitor 1C (CDKN1C), also known as p57KIP2, is a tight-binding, strong inhibitor of several G1 cyclin/CDK complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndrome, a cancer-predisposing syndrome, suggesting that this gene is a tumor suppressor candidate. CDKN1C is widely expressed during organogenesis with its expression declining after maturity. It is part of the CIP/KIP family of cyclin-dependent inhibitors (CKIs) which also includes CDKN1A and CDKN1B.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CDKN1C
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty