Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDIP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AJ5
10802-294EA 423.98 USD InStock
10802-294
Anti-CDIP1 Rabbit Polyclonal Antibody
Antibodies
The p53 tumor-suppressor gene integrates numerous signals that control cell life and death; loss of its functions contributes to the development of most cancers. CDIP is a novel pro-apoptotic target gene whose inhibition abrogates p53-mediated apoptotic responses. Overexpression of CDIP induced apoptosis in transfected cells while siRNA suppression of caspase-8 mRNA blocked this CDIP-induced apoptosis, indicating that the CDIP-dependent apoptosis pathway proceeds through extrinsic cell death pathway. CDIP may thus represent a novel target for drug design to maximize p53 response and sensitize tumor cells to cancer therapy. Multiple isoforms of CDIP are known to exist.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 2.5 µg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CDIP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty