Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD81 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AJ2
10800-706EA 423.98 USD InStock
10800-706
Anti-CD81 Rabbit Polyclonal Antibody
Antibodies
CD81 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family, a group of cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that associates with integrins and appears to promote muscle cell fusion and support myotube maintenance. Along with SCARB1, LDL-R, and CLDN1, CD81 has been reported to be an entry factor for the Hepatitis C virus. Finally, recent evidence indicates that the CD81 gene is localized in a tumor-suppressor gene region and is thus a candidate gene for malignancies.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CD81
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty