Anti-CFTR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AH7
10800-774EA 522.84 USD
10800-774
Anti-CFTR Rabbit Polyclonal Antibody
Antibodies
The cystic fibrosis transmembrane conductance regulator (CFTR) protein is a member of the ATP-binding cassette (ABC) transporter superfamily, and a member of the MRP subfamily that is involved in multi-drug resistance (1,2). CFTR functions as a chloride channel and controls the regulation of other transport pathways (3). Mutations in this gene are associated with the autosomal recessive disorder cystic fibrosis, the most common, fatal, inherited disease of caucasian populations (1).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CFTR
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR