Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDX2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AH2
10800-760EA 423.98 USD InStock
10800-760
Anti-CDX2 Rabbit Polyclonal Antibody
Antibodies
CDX2 is a member of the caudal homeobox family of transcription factors that also includes the proteins CDX1 and CDX4. CDX2 is strongly expressed in the trophectoderm in the blastocyst with expression maintained within the proliferating extra-embryonic ectoderm during development. It is required for the repression of the transcription of the POU5F1/Oct4 and NANOG genes in the outer cells of the blastocyst and is thus essential for the segregation of the inner cell mass and trophectoderm at the blastocyst stage. Aberrant expression of CDX2 is associated with numerous types of carcinomas, including intestinal metaplasia, gastric carcinoma, and acute myeloid leukemia due to deregulated HOX gene expression.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CDX2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty