Anti-CD160 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AG7
10800-668EA 518.29 USD
10800-668
Anti-CD160 Rabbit Polyclonal Antibody
Antibodies
CD160, also known as BY55, is a lipid-anchored cell membrane glycoprotein that contains one immunoglobulin-like domain (1). It is expressed in small intestine, spleen and functional NK and T cytotoxic lymphocytes (1,2). CD160 exists as a disulfide-linked homomultimer that functions as a receptor for MHC (major histocompatability complex) molecules and is thought to regulate the function of NK cells (2,3). Additionally, CD160 interacts with TNFRSF14 and, via this interaction, is able to negatively regulate CD4+ T cell activation, indicating a role in immune system regulation (4).

Recommended Dilutions: ELISA: User Optimized; Immunohistochemsitry: User Optimized; Western Blot: User Optimized; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CD160
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR