Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCP110 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AG2
10800-650EA 423.98 USD InStock
10800-650
Anti-CCP110 Rabbit Polyclonal Antibody
Antibodies
CCP110 is a centrosomal protein required for the centrosome to function as a microtubule organizing center and plays an essential role in centrosome duplication. It is a target of cyclin-dependent kinases (CDKs); disruption of CCP110 phosphorylation leads to unscheduled centrosome separation and polyploidy. CCP110 is also a target of SCF (Cyclin F) ubiquitin ligase complex, leading to its degradation, suggesting that this complex controls centrosome homeostasis and mitotic fidelity through CCP110 degradation. The centriolar kinesin Kif24 also interacts with CP110 and remodel microtubules and regulate ciliogenesis.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 5 µg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CCP110
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty