Anti-CCDC141 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AF2
10800-630EA 518.29 USD
10800-630
Anti-CCDC141 Rabbit Polyclonal Antibody
Antibodies
Coiled-coil domain-containing protein 141 (CCDC141), also known as CAMDI, is a recently identified disrupted in schizophrenia 1 (DISC1)-binding protein that also associates with myosin II. CCDC141 preferentially associates with phosphomyosin II and causes an accumulation of phosphomyosin II at the centrosome in a DISC1-dependent manner. Knockdown of CCDC141 expression by RNAi led to severely impaired radial migration with disoriented chromosomes, suggesting that CCDC141 is required for radial migration through DISC1 and myosin II-mediated centrosome positioning during neuronal development.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CCDC141
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR