Anti-CASR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AE5
10800-568EA 518.29 USD
10800-568
Anti-CASR Rabbit Polyclonal Antibody
Antibodies
The calcium-sensing receptor protein (CASR) is a G protein-coupled receptor that is expressed in the parathyroid hormone (PTH)-producing chief cells of the parathyroid gland, and the cells lining the kidney tubule (1). It senses small changes in circulating calcium concentration and couples this information to intracellular signaling pathways that modify PTH secretion or renal cation handling, thus this protein plays an essential role in maintaining mineral ion homeostasis (1). Mutations in this gene cause familial hypocalciuric hypercalcemia, familial, isolated hypoparathyroidism, and neonatal severe primary hyperparathyroidism (1,2). Recent evidence suggests that activated CASR contributes to the cytokine secretion through the partial MAPK and NF-kappaB pathways in T cells (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CASR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR