Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AD8
10802-282EA 423.98 USD InStock
10802-282
Anti-CASP6 Rabbit Polyclonal Antibody
Antibodies
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. The apoptotic subfamily can be further divided into initiator caspases, which are activated in response to death signals, and executioner caspases, which are activated by the initiator caspases and are responsible for cleavage of cellular substrates that ultimately lead to cell death. Caspase-6 is an executioner caspase that was idientifed based on its homology with human caspases 2 and 3 as well as the C. elegans cell death protein CED-3. It possesses two isoforms, of which only the longer form possesses protease activity. Caspase-6 is highly expressed in adult brain and may play a role in several neuronal pathologies.

Recommended Dilutions: ELISA: 1:5000-1:20,000; Western Blot: 0.5-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty