Anti-CASKIN2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AC3
10800-540EA 518.29 USD
10800-540
Anti-CASKIN2 Rabbit Polyclonal Antibody
Antibodies
Calcium/calmodulin-dependent serine protein kinase (CASK) is a conserved multi-domain scaffolding protein involved in brain development, synapse formation, and establishment of cell polarity. CASKINs (CASK interacting protein ) interact with CASK and is thought to play a role in CASK function, specifically by coupling CASK to distinct downstream effectors. CASKIN2 is a multidomain protein containing six N-terminal ankyrin repeats, one SH3 domain, and two sterile alpha motif domains followed by a long proline-rich sequence and a short conserved C-terminal domain. It is expressed ubiquitously with highest levels present in fetal and adult liver tissue.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 2.5 µg/mL; Immunofluorescence Microscopy: 5 µg/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CASKIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR