Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CARD14 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AB9
10800-532EA 423.98 USD InStock
10800-532
Anti-CARD14 Rabbit Polyclonal Antibody
Antibodies
CARMA proteins belong to the membrane-associated guan-ylate kinase-like (MAGUK) family of proteins that can function as molecular scaffolds that assist assembly of signal transduction molecules. CARMA1, CARMA2, and CARMA3 share high degrees of sequence and functional homology, but their tissue-specific distribution suggests that they serve distinct biological functions in different cell types. As with CARMA1, the CARD domain of CARMA2 has been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kB activation. When expressed in cells, this protein activated NF-kB and induced the phosphorylation of BCL10 Alternative splicing of CARMA2 results in isoforms that possess differential effects on NF-kB activation and endoplasmic reticulum stress-induced cell death.

Recommended Dilutions: ELISA: User Optimized; Immunohistochemsitry: 5 µg/mL ; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CARD14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty