Anti-CARD9 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AB6
10800-526EA 518.29 USD
10800-526
Anti-CARD9 Rabbit Polyclonal Antibody
Antibodies
Apoptosis is related to many diseases and development. Cell death signals are transduced by death domain (DD), death effector domain (DED), and caspase recruitment domain (CARD) containing molecules. CARD containing proteins include some caspases, Apaf-1, CARD4, IAPs, RICK, ARC, RAIDD, BCL-10, and ASC. A novel CARD-containing protein was recently identified and designated CARD9, which interacts with the CARD activation domain of BCL-10. CARD9 associates with BCL-10 and forms a complex within cells. CARD9 induces apoptosis and activates NF-kB. CARD9 is an upstream activator of BCL-10 and NF-kB signaling.

Recommended Dilutions: ELISA: 1:25,000; Western Blot: 2.5 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CARD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR