Anti-CARD10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AB4
10800-522EA 518.29 USD
10800-522
Anti-CARD10 Rabbit Polyclonal Antibody
Antibodies
Apoptosis is related to many diseases and development. Cell death signals are transduced by death domain (DD), death effector domain (DED), and caspase recruitment domain (CARD) containing molecules. CARD containing proteins include some caspases, Apaf-1, CARD4, IAPs, RICK, ARC, RAIDD, Bcl-10, and ASC. A novel CARD-containing protein was recently identified and designated CARD10. This protein belongs to the membrane-associated guanylate kinase-like (MAGUK) family of proteins that can function as molecular scaffolds that assist assembly of signal transduction molecules. CARD 10 interacts with Bcl-10, a Bcl protein that promotes apoptosis, caspase-9 maturation and activation of NF-kB. CARD 10 and the related protein CARMA1 also associate with NEMO, the regulatory subunit of the IkK complex, demonstrating its importance in the regulation of NF-kB transcription factor activation.

Recommended Dilutions: ELISA: 1:50,000-1:100,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 5-10 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CARD10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR