Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CADM3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

600-401-AA6
10802-076EA 423.98 USD InStock
10802-076
Anti-CADM3 Rabbit Polyclonal Antibody
Antibodies
Cell adhesion molecule 3 (CADM3) is a member of a family of immunoglobulin-like, calcium-independent cell-cell adhesion molecules (1,2). CADM3 localizes to non-junctional contact sites of presynaptic nerve terminals, axons and glia cell processes and is thought to play an important role in the formation of synapses, axon bundles and myelinated axons (3). Recently, CADM3 has been shown to suppress the growth and tumorigenic ability of colon cancer cells, indicating that it can act as a tumor suppressor (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CADM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty