Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: 5D7]

Supplier: Rockland Immunochemical

600-301-G94
10801-110EA 423.98 USD InStock
10801-110
Anti-TNFRSF10D Mouse Monoclonal Antibody [clone: 5D7]
Antibodies
DC-SIGN Monoclonal antibody detects human DC-SIGN. Dendritic cells (DCs) that control immune responses were recently found to capture and transport HIV from the mucosal area to remote lymph nodes , where DCs hand over HIV to CD4+ T lymphocytes. DCs also amplify the amount of virus and extend the duration of viral infectivity. Multiple strains of HIV-1, HIV-2 and SIV bind to DCs via DC-SIGN. ICAM-3 is the natural ligand for DC-SIGN. A DC-SIGN homologue (termed DC-SIGNR, L-SIGN, and DC-SIGN2) was identified recently. DC-SIGN forms a novel gene family with DC-SIGNR and many alternatively spliced isoforms of DC-SIGN and DC-SIGNR. The expression of DC-SIGN was found in mucosal tissues including placenta, small intestine, and rectum. Anti-DC-SIGN Monoclonal antibodies are ideal for investigators involved in infectious disease research.

Recommended Dilutions: Western Blot: 1-2 ug/mL; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: TNFRSF10D
Clonality: Monoclonal
Clone: 5D7
Conjugation: Unconjugated
Epitope: Extracellular
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty