Anti-ATP1A1 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-9155 200-9155
10800-236EA 1289.92 USD
10800-236 CA10800-236
Anti-ATP1A1 Chicken Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:5,000 - 1:20,000; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:5,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: ATP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR