Anti-ANKLE1 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-Y16 200-901-Y16
10800-024EA 518.29 USD
CA10800-024 10800-024
Anti-ANKLE1 Chicken Polyclonal Antibody
Antibodies
Ankyrins are membrane adaptor molecules that play important roles in coupling integral membrane proteins to the spectrin-based cytoskeleton network. ANKLE1 (Ankyrin repeat and LEM domain containing 1) is a member of ankyrin repeat and LEM domain containing family. It is a potential single-pass membrane protein that contains ankyrin repeats and a Lem domain. Ankyrin repeats mediate protein-protein interactions and are among the most common structural motifs in known proteins. While the function of this protein is still under investigation, its gene locus has been associated with susceptibility to ovarian and breast cancer.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ANKLE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR