Anti-TUBA3C Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-Y06 200-901-Y06
10799-980EA 534.3 USD
10799-980 CA10799-980
Anti-TUBA3C Chicken Polyclonal Antibody
Antibodies
Alpha-tubulin belongs to the tubulin superfamily, which is composed of six distinct families. Along with beta-tubulins, alpha-tubulins are the major components of microtubules. These microtubules are involved in a wide variety of cellular activities ranging from mitosis and transport events to cell movement and the maintenance of cell shape. Alpha- and beta-tubulin dimers are assembled to 13 protofilaments that form a microtubule of 22-nm diameter. Tyrosine ligase adds a C-terminal tyrosine to monomeric alpha-tubulin. Assembled microtubules can again be detyrosinated by a cytoskeleton-associated carboxypeptidase. Another post-translational modification of detyrosinated alpha-tubulin is C-terminal polyglutamylation, which is characteristic of microtubules in neuronal cells and the mitotic spindle. Like GAPDH and beta-Actin, this antibody makes an excellent loading control in immunoblots.

Recommended Dilutions: ELISA: 1:5000-1:10000; Western Blot: 0.5 - 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TUBA3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR