Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADAMTSL5 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-X69 200-901-X69
10799-574EA 423.98 USD InStock
CA10799-574 10799-574
Anti-ADAMTSL5 Chicken Polyclonal Antibody
Antibodies
ADAMTSL5 belongs to a large superfamily containing 20 ADAMTS proteases and at least eight ADAMTS-like proteins. ADAMTS proteases are secreted enzymes with a conserved organization that includes a metalloprotease domain and an ancillary domain containing one or more thrombospondin type 1 repeats (TSR). The ADAMTS-like subfamily comprises proteins homologous to the ADAMTS ancillary domains but lacking the protease domain and hence lacking catalytic activity. The exact role of ADAMTSL5 is currently unknown, but other ADAMTS-like proteins appear to have regulatory roles in the extracellular matrix.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 μg/mL; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ADAMTSL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty