Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAP43 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-X10
10801-540EA 419.61 USD InStock
10801-540
Anti-GAP43 Chicken Polyclonal Antibody
Antibodies
GAP-43 is thought to have an important role in development and plasticity because it is expressed at high levels in neuronal growth cones during development and during axonal regeneration. There is also evidence from knockout animals that GAP-43 serves to amplify pathfinding signals from the growth cone. GAP-43 is thought to mediate at least some of these effects via interaction with actin. Importantly, phosphorylation at Ser41 by protein kinase C modulates the interaction of GAP-43 with actin and may also affect neurotransmitter release during forms of plasticity like LTP. GAP-43 Antibody is ideal for researchers interested in neuroscience research.

Recommended Dilutions: ELISA: 1:20,000; Immunofluorescence Microscopy: 1:500; Western Blot: 1:2000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Chicken
Isotype:
Reactivity: Rat, Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty