Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FCHSD1 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-BE0
10802-758EA 423.98 USD InStock
10802-758
Anti-FCHSD1 Chicken Polyclonal Antibody
Antibodies
The FCH and double SH3 domains protein 1 (FCHSD1, also known as NWK2) and the related protein FCHSD2 were initially identified in silico as distantly related proteins to FNBP1 and FNBP2. Both share the common domain structure consisting of FCH, FBH, two SH3 and C-terminal proline-rich domains. While little is known of the function of FCHSD1, the related protein NWK is an adaptor protein that is thought to regulate Rho activity downstream of Robo receptors, suggesting that FCHSD1 may be involved in synaptic morphology by regulating actin dynamics in presynaptic terminals.

Recommended Dilutions: ELISA: 1:5000-1:10000; Western Blot: 0.5 - 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FCHSD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty