Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RMDN2 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-BD6
10801-336EA 423.98 USD InStock
10801-336
Anti-RMDN2 Chicken Polyclonal Antibody
Antibodies
The human protein FAM82A1, also known as regulator of microtubule dynamics 2 (RMD2), was one of three proteins identified as related to a family of microtubule-associated proteins in C. elegans. FAM82A1 contains multiple coiled-coil domains and localize to the spindle microtubules and spindle poles during cell division. During interphase, RMD2 localizes to the cytoplasm with protein observed in the microtubule lattice and perinuclear region.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 1-2 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: RMDN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty