Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPINK7 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-AY8
10802-566EA 423.98 USD InStock
10802-566
Anti-SPINK7 Chicken Polyclonal Antibody
Antibodies
The esophageal cancer-susceptibility gene 2 (ECRG2), also known as SPINK7, is a novel tumor suppressor gene identified from the human esophagus. It interacts directly with metallothionein 2A and urokinase-type plasminogen activator (uPA), and downregulates the activity of uPA, leading to reduced cancer cell migration, invasion and metastasis. ECRG2 forms a complex with uPA and its receptor uPAR, modifying the dynamic association of uPAR with beta1 integrins and disrupting the Src/MAP kinase pathway that normally stimulates cell migration and invasion. ECRG2 may thus represent a novel therapeutic target for cancer.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 μg/mL; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: SPINK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty