Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMPRSS11A Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-AY7
10802-564EA 423.98 USD InStock
10802-564
Anti-TMPRSS11A Chicken Polyclonal Antibody
Antibodies
Epidermal type-II transmembrane serine protease (ECRG1), also known as TMPRSS11A, is a member of a novel esophageal cancer-related gene family. It is a type-II transmembrane serine protease which may play a role in cellular senescence. It is expressed in esophagus, liver, colon and lung. Overexpression of ECRG1 inhibits cell growth and induces G1 cell cycle arrest. It is down-regulated in esophagus cancers (EC) and might play important roles in the initiation and progression of EC.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TMPRSS11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty