Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NSRP1 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical

200-901-AF6
10800-638EA 423.98 USD InStock
10800-638
Anti-NSRP1 Chicken Polyclonal Antibody
Antibodies
CCDC55, also known as Nuclear speckle-related protein 70 (NSrp70), was initially identified by the NIH Mammalian Gene Collection. Nuclear speckles are known to be the storage sites of mRNA splicing regulators. CCDC55 has been suggested to modulate alternative pre-mRNA splicing in vivo. It co-localizes and physically interacts with the SR proteins SC35 and ASF/SF2, essential splicing factors which are required for constitutive splicing and can regulate alternative mRNA splicing. Loss of CCDC55 in mice leads to early embryonic lethality highlights the importance of the functional role of CCDC55.

Recommended Dilutions: ELISA: 1:5000-1:10000; Immunofluorescence Microscopy: 20 μg/mL; Western Blot: 0.5-1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: NSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty