Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical

200-4336S
10800-328EA 124.57 USD InStock
10800-328
Anti-GLB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
β-galactosidase, (beta-gal or β-gal), is a hydrolase enzyme that catalyzes the hydrolysis of β-galactosides into monosaccharides. It may also cleave fucosides and arabinosides but with much lower efficiency. It is an essential enzyme in the human body. Deficiencies in the protein can result in galactosialidosis or Morquio B syndrome.

Recommended Dilutions: ELISA: 1:2,000 - 1:5,000; Immunohistochemsitry: 1:250; Western Blot: 1:500 - 1:2,000; 0.01% (w/v) Gentamicin Sulfate. Do NOT add Sodium Azide!

Type: Tag
Antigen: beta Galactosidase
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty