Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-406-N98S 200-406-N98S
10799-926EA 124.57 USD InStock
CA10799-926 10799-926
Anti-AKT3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
AKT is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as AKT1, Protein Kinase B (PKB) and rac (related to A and C kinases). AKT is a key regulator of many signal transduction pathways. AKT Exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. Anti-AKT Antibody is ideal for investigators involved in Cell Signaling, Cancer, Neuroscience, Signal Transduction research.

Recommended Dilutions: ELISA: User Optimized; Flow Cytometry: User Optimized; Immunohistochemsitry: 1:500 - 1:2,000; Immunofluorescence Microscopy: User Optimized; Immunoprecipitation: 1:500 - 1:2,000; Western Blot: 1:500 - 1:2,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty