Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BACE Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

200-401-G83
10801-976EA 423.98 USD InStock
10801-976
Anti-BACE Rabbit Polyclonal Antibody
Antibodies
BAFF antibody detects human BAFF. Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated BAFF (for B cell Activating Factor belonging to the TNF Family), BLyS (for B Lymphocyte Stimulator), TALL-1 (for TNF- and ApoL-related Leukocyte-expressed Ligand), and THANK (for TNF Homologue that Activate Apoptosis, NF-alpha;B and c-jun N-terminal Kinase). BAFF/BLyS was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF and its receptors are essential for B cell development, survival, and humoral immune responses. BAFF is involved in the development of autoimmune diseases including systemic lupus erythaematosus and rheumatoid arthritis. Anti-BAFF antibodies are ideal for investigators involved in NFkappaB, Cytokine and Growth Factor research.

  • Recommended Dilutions: Western Blot: 0.25-1 ug/mL; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: BACE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty