Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFRA2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical

200-401-BK6
10801-686EA 423.98 USD InStock
10801-686
Anti-GFRA2 Rabbit Polyclonal Antibody
Antibodies
Members of the glial cell line-derived neurotrophic factor (GDNF) family, including GDNF and neurturin (NTN) play key roles in the control of vertebrate neuron survival and differentiation. Physiological responses to NTN require the presence of a novel glycosylphosphadidylinositol linked protein NTNRalpha, which is a cell surface receptor for NTN. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently. NTNRalpha was also termed GDNFRb, Ret ligand 2 (RETL2) or TGFbeta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFRa-2 recently. GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 μg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: GFRA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty